Trypan blue 60% (dye content) ≥80% (by HPLC), Sigma-Aldrich®
:72-57-1
:
:C₃₄H₂₄N₆Na₄O₁₄S₄
:MFCD00003969
:960.82 g/mol
:>300 °C (lit.)
:M-Clarity™ quality level = MQ200
:200-786-7
:GHS08
:Room Temperature
- Appearance (Color):Dark Green to Brown or Brown-Black
- Appearance (Form):Powder or Crystals or Granules
- Infrared spectrum:Conforms to Structure
- Wavelength:603 - 607 nm
- Extinction Coefficient, c = 0.01 g/L; Methanol:≥60000
- Carbon:≥24.6 %
- Nitrogen:≥5.0 %
- Purity (HPLC):≥80.0 %
- Solubility (Turbidity); Opaque, c = 1mg/ml; Water:Conforms
- Solubility (Color):Dark Blue
- Recommended Retest Period:5 Years
Frequently Bought Together