
Trypan blue 60% (dye content)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:72-57-1
:
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969
:960.82 g/mol
Merck Index:14,9792
Melting Pt:>300 °C
EINECS:200-786-7
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together