Order Entry
Austria
ContactUsLinkComponent
2-Hydroxy-4-methoxy-5-sulphobenzophenone
2-Hydroxy-4-methoxy-5-sulphobenzophenone
Catalog # MOLE39486176-25G
Supplier:  Molekula
CAS Number:  
2-Hydroxy-4-methoxy-5-sulphobenzophenone
Catalog # MOLE39486176-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :308.30 g/mol
EINECS:223-772-2
CAS Number:4065-45-6
Formula:C₁₄H₁₂O₆S
MDL Number:MFCD00024962