Order Entry
Austria
ContactUsLinkComponent
Trypan blue 4 g/l in aqueous solution cell culture reagent
Health
Trypan blue 4 g/l in aqueous solution cell culture reagent
Catalog # K940-100ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Trypan blue 4 g/l in aqueous solution cell culture reagent
Catalog # K940-100ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Abs.@ Lambda Max (1:100, Water):
    1 - 1.3
  • Lambda Max (1:100, Water):
    585 - 595 nm

Specification Test Results

About this item

 :
 :960.82 g/mol
EINECS:200-786-7
 :GHS08
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969