Order Entry
Austria
ContactUsLinkComponent
Sulphur standard dissolved in oil, 5000 µg/g S in mineral oil for wear metal analysis, Specpure®
Sulphur standard dissolved in oil, 5000 µg/g S in mineral oil for wear metal analysis, Specpure®
Artikelnr. 43870.18
CAS Number:  
Sulphur standard dissolved in oil, 5000 µg/g S in mineral oil for wear metal analysis, Specpure®
Artikelnr. 43870.18
CAS Number:  

Specifications

  • Concentration:
    5000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Description:
    Standard for wear metal analysis
  • MDL No.:
    MFCD00242296
  • Element:
    Sulphur
  • Symbol:
    S

Specifications

About this item

Physical form: liquid
MDL No.: MFCD00242296