Order Entry
Austria
ContactUsLinkComponent
Thallium standard dissolved in oil, 1000 µg/g Tl in mineral oil for wear metal analysis, Specpure®
Thallium standard dissolved in oil, 1000 µg/g Tl in mineral oil for wear metal analysis, Specpure®
Catalog # 45306.18
CAS Number:  
Thallium standard dissolved in oil, 1000 µg/g Tl in mineral oil for wear metal analysis, Specpure®
Catalog # 45306.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Description:
    Standard for wear metal analysis
  • MDL No.:
    MFCD01095325
  • Element:
    Thallium
  • Symbol:
    Tl

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD01095325