Order Entry
Austria
ContactUsLinkComponent
Selenium standard dissolved in oil, 1000 µg/g Se in mineral oil for wear metal analysis, Specpure®
Selenium standard dissolved in oil, 1000 µg/g Se in mineral oil for wear metal analysis, Specpure®
Catalog # 45305.18
CAS Number:  
Selenium standard dissolved in oil, 1000 µg/g Se in mineral oil for wear metal analysis, Specpure®
Catalog # 45305.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Description:
    Standard for wear metal analysis
  • Environmentally Preferable:
  • MDL No.:
    MFCD00134090
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item

Physical form: liquid
MDL No.: MFCD00134090