Order Entry
Austria
ContactUsLinkComponent
Phosphorus standard dissolved in oil, 1,000 µg/g P in mineral oil for wear metal analysis, Specpure®
Phosphorus standard dissolved in oil, 1,000 µg/g P in mineral oil for wear metal analysis, Specpure®
Catalog # 14210.18
CAS Number:  
Phosphorus standard dissolved in oil, 1,000 µg/g P in mineral oil for wear metal analysis, Specpure®
Catalog # 14210.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00133771
  • Element:
    Phosphorus
  • Symbol:
    P

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00133771