Order Entry
Austria
ContactUsLinkComponent
Potassium standard dissolved in oil, 1000 µg/g K in mineral oil for wear metal analysis, Specpure®
Potassium standard dissolved in oil, 1000 µg/g K in mineral oil for wear metal analysis, Specpure®
Artikelnr. 14216.18
CAS Number:  
Potassium standard dissolved in oil, 1000 µg/g K in mineral oil for wear metal analysis, Specpure®
Artikelnr. 14216.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00133776
  • Element:
    Potassium
  • Symbol:
    K

Specifications

About this item

Physical form: liquid
MDL No.: MFCD00133776