Order Entry
Austria
ContactUsLinkComponent
Boron standard dissolved in oil, 1,000 µg/g B in mineral oil for wear metal analysis, Specpure®
Boron standard dissolved in oil, 1,000 µg/g B in mineral oil for wear metal analysis, Specpure®
Catalog # 14227.18
CAS Number:  
Boron standard dissolved in oil, 1,000 µg/g B in mineral oil for wear metal analysis, Specpure®
Catalog # 14227.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00134034
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00134034