Order Entry
Austria
ContactUsLinkComponent
 
Beryllium standard dissolved in oil, 1,000 µg/g Be in Base Oil 20
Catalog # PROOCON150-100-045
CAS Number:  
Beryllium standard dissolved in oil, 1,000 µg/g Be in Base Oil 20
Catalog # PROOCON150-100-045
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Base Oil 20
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Element:
    Beryllium
  • Symbol:
    Be

Specifications

About this item