Order Entry
Austria
ContactUsLinkComponent
Strontium standard dissolved in oil, 1,000 µg/g Sr in mineral oil for wear metal analysis, Specpure®
Strontium standard dissolved in oil, 1,000 µg/g Sr in mineral oil for wear metal analysis, Specpure®
  14205.18
 :  
Strontium standard dissolved in oil, 1,000 µg/g Sr in mineral oil for wear metal analysis, Specpure®
  14205.18
 :  

 

  • Concentration:
    1000 µg/g
  • Matrix:
    oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Description:
    Standard for wear metal analysis
  • MDL No.:
    MFCD00134060
  • Element:
    Strontium
  • Symbol:
    Sr

 

 

Physical Form: Liquid
MDL No.: MFCD00134060