Order Entry
Austria
ContactUsLinkComponent
Mercury standard dissolved in oil, 1,000 µg/g Hg in mineral oil for wear metal analysis, Specpure®
Mercury standard dissolved in oil, 1,000 µg/g Hg in mineral oil for wear metal analysis, Specpure®
Catalog # 43756.18
CAS Number:  
Mercury standard dissolved in oil, 1,000 µg/g Hg in mineral oil for wear metal analysis, Specpure®
Catalog # 43756.18
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00011035
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00011035