Molybdenum standard dissolved in oil, 5000 µg/g Mo in mineral oil for wear metal analysis, Specpure®
Catalog # 42445.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:Mineral oil
- Pack type:Glass bottle for solids
- Pk:50 g
- MDL No.:MFCD00003465
- Element:Molybdenum
- Symbol:Mo
Specifications
About this item
Physical Form: Liquid
MDL No.: MFCD00003465