Molybdenum standard dissolved in oil, 1000 µg/g Mo in mineral oil for wear metal analysis, Specpure®
14213.18
:
- Concentration:1000 mg/L
- Matrix:Mineral oil
- Pack type:Glass bottle for solids
- Pk:50 g
- MDL No.:MFCD00003465
- Element:Molybdenum
- Symbol:Mo
Physical Form: Liquid
MDL No.: MFCD00003465