Order Entry
Austria
ContactUsLinkComponent
4-Bromo-3-methyl-1H-pyrazole ≥97%
Irritant
4-Bromo-3-methyl-1H-pyrazole ≥97%
Catalog # B20069.06
CAS Number:  
Irritant
4-Bromo-3-methyl-1H-pyrazole ≥97%
Catalog # B20069.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:13808-64-5
 :
Formula:C₄H₅BrN₂
MDL Number:MFCD00005241
 :161.01 g/mol
Melting Pt:73…77 °C
EINECS:237-456-7
 :GHS07
Storage Temperature:Ambient