Order Entry
Austria
ContactUsLinkComponent
Lead standard dissolved in oil, 1000 µg/g Pb in mineral oil for wear metal analysis, Specpure®
Lead standard dissolved in oil, 1000 µg/g Pb in mineral oil for wear metal analysis, Specpure®
Catalog # 14208.18
CAS Number:  
Lead standard dissolved in oil, 1000 µg/g Pb in mineral oil for wear metal analysis, Specpure®
Catalog # 14208.18
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00134050
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item

Physical form: liquid
MDL No.: MFCD00134050