Order Entry
Austria
ContactUsLinkComponent
Chlorfenapyr
Chlorfenapyr
Catalog # EHERC11247500
Supplier:  EHRENSTORFER
CAS Number:  
Chlorfenapyr
Catalog # EHERC11247500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
CAS Number:122453-73-0
Formula:C₁₅H₁₁BrClF₃N₂O
MDL Number:MFCD01631152