2-Sulphobenzoic cyclic anhydride ≥90%, Sigma-Aldrich®
Catalog # 191698-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance (Color):White to Grey and Faint, Beige to Beige
- Appearance (Form):Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
- Titration with NaOH:89.5 - 110.5 %
- Infrared Spectrum:Conforms to Structure
Specification Test Results
About this item
:
:184.17 g/mol
Boiling Pt:184…186 °C/18 mmHg (lit.)
Melting Pt:116 °C
:M-Clarity™ quality level = MQ200
EINECS:201-322-6
:GHS07
CAS Number:81-08-3
Formula:C₇H₄O₄S
MDL Number:MFCD00005879