Folic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
CAS Number:59-30-3
:
Formula:C₁₉H₁₉N₇O₆
MDL Number:MFCD00079305
Molecular Weight:441.41 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-419-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together