Buffer, TRIS solution pH 8,0 (1 mol/L), Sigma-Aldrich®
Supplier: Merck
Certificates
Specifications
- M-Clarity quality level:MQ200
Specifications
Frequently Bought Together
Short-term tubes for accuro®
Supplier: Dräger
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Laboratory trays, LaboPlast®
Supplier: BURKLE
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nonidet® P 40 Substitute (NP-40), Millipore®
Supplier: Merck Millipore (Calbiochem)
Ethyl acetate ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals