Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 8 (1 mol/L)

Buffer, TRIS solution pH 8 (1 mol/L)

TRIS buffer solution (1 mol/l), 8.0, Glass bottle, 1 L
Catalog # A4577.1000
Supplier:   PanReac AppliChem
Buffer, TRIS solution pH 8 (1 mol/L)
Catalog # A4577.1000
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TRIS solution pH 8 (1 mol/L)

TRIS buffer solution (1 mol/l), 8.0, Glass bottle, 1 L

CAS Number:  
Price
54,80 €
Log in for availability
1 * 1 L
Product Details & Documents
  • DNases/RNases/Proteases: not detectable
  • pH (20°C; adjusted with HCl): 8.0 ± 0.1
  • Composition: Tris: 121.14 g/L (1 M)

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS buffer solution (1 mol/l)
  • pH value:
    8.0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.