Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7,5 (1 mol/L)

Buffer, TRIS solution pH 7,5 (1 mol/L)

TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 500 mL
Catalog # A4263.0500
Supplier:   PanReac AppliChem
Buffer, TRIS solution pH 7,5 (1 mol/L)
Catalog # A4263.0500
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TRIS solution pH 7,5 (1 mol/L)

TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
38,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TRIS buffer solution pH 7,5 (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.