Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Biological Buffers
VWR® Tris Buffer Solution, pH 7,5 (1 mol/L), UltraPure Grade
VWR® Tris Buffer Solution, pH 7,5 (1 mol/L), UltraPure Grade
VWR® Tris Buffer Solution, pH 7,5 (1 mol/L), UltraPure Grade
Catalog # E691-500ML
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
VWR® Tris Buffer Solution, pH 7,5 (1 mol/L), UltraPure Grade
Catalog # E691-500ML
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
500 mL
Description:
TRIS buffer solution pH 7,5 (1 mol/l)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation