Buffers, PIPES, 0,5 M solutions
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
: Storage Temperature: 2 to 8 °C
- CAS No.:5625-37-6
- MDL No.:MFCD00006159
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- Appearance:Colourless
Frequently Bought Together