Order Entry
Austria
ContactUsLinkComponent
Buffers, Sodium Citrate, 0,5M Solutions
Buffers, Sodium Citrate, 0,5M Solutions
Certificates
Buffers, Sodium Citrate, 0,5M Solutions
Certificates

Specifications

  • MDL No.:
    MFCD00012462
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless

Specifications

Frequently Bought Together