Buffer, potassium acetate, 1 M aq, soln, pH 7,5, RNAse free
Catalog # J60832.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- CAS No.:127-08-2
- Description:Potassium acetate, 1M aq, solution, pH 7,5, RNAse free
- MDL No.:MFCD00012458
- Physical form:Liquid
- Appearance:Colourless
Specifications
About this item
CAS No.: 127-08-2
Molecular formula: C₂H₃KO₂
Formula weight: 98,15
Physical form: liquid
Appearance: colourless