VWR® Buffer, HEPES Solution pH 7,3 (1 mol/L)
Supplier: VWR Chemicals
Certificates
Frequently Bought Together
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Freezing containers, Mr. Frosty, Nalgene®
Supplier: Thermo Fisher Scientific
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Restore™ Western Blot Stripping Buffer
Supplier: Thermo Fisher Scientific
HEPES Buffer, 1 M Sterile, Cell Culture Grade, MP Biomedicals
Supplier: MP Biomedicals