Order Entry
Austria
ContactUsLinkComponent
Buffer, HEPES solution (1 mol/L), Corning®
Buffer, HEPES solution (1 mol/L), Corning®
Catalog # 25-060-CI
Supplier:  Corning
CAS Number:  
undefined
Buffer, HEPES solution (1 mol/L), Corning®
Catalog # 25-060-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    6
  • Description:
    HEPES buffer solution (1 mol/l)
  • Storage temperature:
    Ambient

Specifications

About this item

HEPES (N-2-hydroxyethylpiperazine-N'-2-ethanesulphonic acid) provides an alternative to the use of bicarbonate for buffering because it does not require elevated levels of CO2

Unlike the low pKa of bicarbonate, HEPES has a pKa of 7.66 making it a much stronger buffer in the pH 7.2-7.6 range than bicarbonate buffer. Potential disadvantages of HEPES include toxicity over 100 mM and lack of nutritive value to the cell.