Organic reference standard, Musk ketone 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERLA15359000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:81-14-1
- Description:Musk ketone 10 µg/ml in cyclohexane
- Formula:C₁₄H₁₈N₂O₅
- Description:Musk ketone