Ethyl trimethylacetopyruvate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13395-36-3
:
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00052319
:200.24 g/mol
Boiling Pt:68…70 °C (0.5 mmHg)
Storage Temperature:Ambient
EINECS:236-478-4
Related Information
Frequently Bought Together