Order Entry
Austria
ContactUsLinkComponent
Calcium pyruvate
Calcium pyruvate
Catalog # MOLE19918840-100G
Supplier:  Molekula
CAS Number:  
Calcium pyruvate
Catalog # MOLE19918840-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Calcium 2-oxopropanoate
 :
 :214.19 g/mol
EINECS:257-599-9
CAS Number:52009-14-0
Formula:C₆H₆CaO₆
MDL Number:MFCD00037199