Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Thiodicarb

Catalog #
Thiodicarb
Catalog #
Supplier:  
CAS Number:  59669-26-0
Thiodicarb
CAS Number:  59669-26-0
Product Details & Documents
  • CAS Number:59669-26-0
  • Formula:C₁₀H₁₈N₄O₄S₃
  • MDL Number:MFCD00145401
  • EINECS:261-848-7
  • UN:2811
  • ADR:6.1,I