Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Chlordane
cis-Chlordane
cis-Chlordane
cis-Chlordane
Catalog # PROONMIAP1624
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
cis-Chlordane
Catalog # PROONMIAP1624
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
EINECS:
225-825-5
CAS Number:
5103-71-9
Formula:
C₁₀H₆Cl₈
Product Details