Desiccators, with porcelain plate
Supplier: Kavalier
Certificates
About this item
Simax borosilicate glass, novus pattern.
- Vacuum or knob lid type with tubular in centre of lid
- Cover ground to fit
- Spare lids are available for these desiccators
Delivery: Supplied with lid, removable stopcock (vacuum type only) and perforated ceramic disc.