Order Entry
Austria
ContactUsLinkComponent
Tissue DNA kit, genomic DNA isolation from tissue, E-Z 96®
Tissue DNA kit, genomic DNA isolation from tissue, E-Z 96®
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Tissue DNA kit, genomic DNA isolation from tissue, E-Z 96®
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Sample size:
    30 mg
  • Description:
    E-Z 96® Tissue DNA Kit

Specifications

Related Information

Frequently Bought Together