Order Entry
Austria
ContactUsLinkComponent
 
Plasmid isolation, single-strand phage DNA, M-13 isolation kits, E.Z.N.A.® and E-Z 96®
 
Plasmid isolation, single-strand phage DNA, M-13 isolation kits, E.Z.N.A.® and E-Z 96®

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    ssDNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Sample size:
    1 - 3 ml
  • Description:
    E.Z.N.A.® M-13 DNA Mini Kit

 

Frequently Bought Together