Order Entry
Austria
ContactUsLinkComponent
Tubing connectors, Y-shaped, PP
Tubing connectors, Y-shaped, PP
Catalog # SEMA4660
Supplier:  SEMADENI
undefined
Tubing connectors, Y-shaped, PP
Catalog # SEMA4660
Supplier:  SEMADENI

Specifications

  • Hole Ø:
    5,0 mm
  • Tubing int.Ø:
    6 - 9 mm
  • Pk:
    1

Specifications

About this item

Connectors, Y- shaped, made of PP, natural color.

  • Strong design with long, conical connections for large tolerance ranges
  • Temperature resistant up to 120 °C, sterilisable