Tubing connectors, straight
: SEMADENI
Connectors, made of white PP, with tapered ends, for connecting tubing with the same diameter.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
Frequently Bought Together