Orders LinkContactUsLinkComponent
Empty solid phase extraction columns, CHROMABOND®
Supplier:   MACHEREY-NAGEL
Empty solid phase extraction columns, CHROMABOND®
Supplier:  MACHEREY-NAGEL
Zertifikate
Empty solid phase extraction columns, CHROMABOND®
Empty solid phase extraction columns, CHROMABOND®
14 Optionen unten verfügbar
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
  • Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
  • Reservoir columns from PP for application of medium-size samples
  • Accessories for SPE columns (filter elements, adapters)
Product Family Options
CHROMABOND® empty columns
Produktinformationen
  • Capacity
    Availability
    Price
  • MANA730159
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    1 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 200,00 €
    200,00 €
    1 * 100 ST
       200,00 €
  • MANA730160
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    3 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 50 ST
    1 * 50 ST - 129,00 €
    129,00 €
    1 * 50 ST
       129,00 €
  • MANA730161
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    6 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 30 ST
    1 * 30 ST - 88,90 €
    88,90 €
    1 * 30 ST
       88,90 €
  • MANA730230
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    15 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 20 ST
    1 * 20 ST - 105,00 €
    105,00 €
    1 * 20 ST
       105,00 €
  • MANA730380
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    30 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 20 ST
    1 * 20 ST - 111,00 €
    111,00 €
    1 * 20 ST
       111,00 €
  • MANA730355
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    45 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 20 ST
    1 * 20 ST - 145,00 €
    145,00 €
    1 * 20 ST
       145,00 €
  • MANA730158
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    70 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 20 ST
    1 * 20 ST - 157,00 €
    157,00 €
    1 * 20 ST
       157,00 €
  • MANA730474
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    Empty PP columns with PE frits, one filter element is already inserted in the PP column
    150 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 20 ST
    1 * 20 ST - 172,00 €
    172,00 €
    1 * 20 ST
       172,00 €
  • MANA730171
    Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
    Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
    3 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 50 ST
    1 * 50 ST - 255,00 €
    255,00 €
    1 * 50 ST
       255,00 €
  • MANA730172
    Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
    Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
    6 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 30 ST
    1 * 30 ST - 200,00 €
    200,00 €
    1 * 30 ST
       200,00 €
  • MANA732501
    Empty LV PP columns with PE frits, for 200/500 mg adsorbent weight
    Empty LV PP columns with PE frits, for 200/500 mg adsorbent weight
    15 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 50 ST
    1 * 50 ST - 202,00 €
    202,00 €
    1 * 50 ST
       202,00 €
Reservoir columns for application of medium-size samples
Produktinformationen
  • Capacity
    Availability
    Price
  • MANA730103
    Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
    Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
    30 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 10 ST
    1 * 10 ST - 114,00 €
    114,00 €
    1 * 10 ST
       114,00 €
  • MANA730382
    Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
    Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
    70 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 10 ST
    1 * 10 ST - 121,00 €
    121,00 €
    1 * 10 ST
       121,00 €
  • MANA730389
    Reservoir columns, PP, with one adapter for 15, 45, 70 ml CHROMABOND® PP columns
    Reservoir columns, PP, with one adapter for 15, 45, 70 ml CHROMABOND® PP columns
    70 ml
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 10 ST
    1 * 10 ST - 166,00 €
    166,00 €
    1 * 10 ST
       166,00 €
Accessories
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    MANA732019
    Einloggen für Verfügbarkeit
    1 * 250 ST - 132,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    MANA732020
    Einloggen für Verfügbarkeit
    1 * 250 ST - 185,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Adapters (PP) for PP columns (1, 3 and 6 ml)
    Adapters (PP) for PP columns (1, 3 and 6 ml)
    MANA730101
    Einloggen für Verfügbarkeit
    1 * 10 ST - 96,10 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Adapters (PVDF) for glass columns (3 and 6 ml)
    Adapters (PVDF) for glass columns (3 and 6 ml)
    MANA730105
    Einloggen für Verfügbarkeit
    1 * 10 ST - 200,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 70 ml
    PE frits for PP columns, 70 ml
    MANA730026
    Einloggen für Verfügbarkeit
    1 * 250 ST - 281,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Glass fiber frits for glass columns, 6 ml
    Glass fiber frits for glass columns, 6 ml
    MANA730192
    Einloggen für Verfügbarkeit
    1 * 250 ST - 238,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 15 ml
    PE frits for PP columns, 15 ml
    MANA730351
    Einloggen für Verfügbarkeit
    1 * 250 ST - 238,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 45 ml
    PE frits for PP columns, 45 ml
    MANA730356
    Einloggen für Verfügbarkeit
    1 * 250 ST - 273,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 150 ml
    PE frits for PP columns, 150 ml
    MANA730475
    Einloggen für Verfügbarkeit
    1 * 250 ST - 302,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 30 ml
    PE frits for PP columns, 30 ml
    MANA730034
    Einloggen für Verfügbarkeit
    1 * 250 ST - 258,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Adapters (PE) for PP columns (30 and 70 ml)
    Adapters (PE) for PP columns (30 and 70 ml)
    MANA730566
    Einloggen für Verfügbarkeit
    1 * 1 KIT - 29,10 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Adapters (PE) for PP columns (15, 45 and 70 ml)
    Adapters (PE) for PP columns (15, 45 and 70 ml)
    MANA730385
    Einloggen für Verfügbarkeit
    1 * 10 ST - 125,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • Glass fiber frits for glass columns, 3 ml
    Glass fiber frits for glass columns, 3 ml
    MANA730191
    Einloggen für Verfügbarkeit
    1 * 250 ST - 196,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 1 ml
    PE frits for PP columns, 1 ml
    MANA730164
    Einloggen für Verfügbarkeit
    1 * 250 ST - 130,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 6 ml
    PE frits for PP columns, 6 ml
    MANA730163
    Einloggen für Verfügbarkeit
    1 * 250 ST - 226,00 €
Produktinformationen
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 3 ml
    PE frits for PP columns, 3 ml
    MANA730162
    Einloggen für Verfügbarkeit
    1 * 250 ST - 184,00 €