Culture tubes, rimless
Supplier: BELLCO GLASS
Certificates
About this item
Manufactured from type 1 class A borosilicate glass with plain top.
- Very good chemical resistance
- Very good temperature resistance
Caps, stainless steel, without fingers
Product Information
Frequently Bought Together