Orders LinkContactUsLinkComponent
Erlenmeyer flasks, graduated, with screw cap
Marketsource
Supplier:   BELLCO GLASS
Erlenmeyer flasks, graduated, with screw cap
Supplier:  BELLCO GLASS
Marketsource
Certificates
Erlenmeyer flasks, graduated, with screw cap
Erlenmeyer flasks, graduated, with screw cap
4 Options Available Below
Product Details & Documents
Manufactured from type 1 class A borosilicate glass, these flasks are ideal for media storage and mixing. Long-skirted phenolic rubber lined screw cap protects flask contents from contamination.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Graduated to approximate volumes
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 25 ml
    65 mm
    41 mm
    18-415
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 1 210,00 €
    1 210,00 €
    1 * 24 ST
       1 210,00 €
    Specifications:
    Marketsource
    Marketsource
  • 50 ml
    85 mm
    50 mm
    24-415
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 1 130,00 €
    1 130,00 €
    1 * 24 ST
       1 130,00 €
    Specifications:
    Marketsource
    Marketsource
  • 2000 ml
    275 mm
    158 mm
    38-415
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 600,00 €
    600,00 €
    1 * 3 ST
       600,00 €
    Specifications:
    Marketsource
    Marketsource
  • 3000 ml
    320 mm
    184 mm
    38-415
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 776,00 €
    776,00 €
    1 * 3 ST
       776,00 €
    Specifications:
    Marketsource
    Marketsource
Screw caps
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Black
    Screw caps, deep skirted, with white rubber liner
    15-415
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 134,00 €
    134,00 €
    1 * 144 ST
       134,00 €
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Black
    Screw caps, deep skirted, with white rubber liner
    38-415
    Log in for availability
    Log in for availability
    1 * 36 ST
    1 * 36 ST - 108,00 €
    108,00 €
    1 * 36 ST
       108,00 €
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Black
    Screw caps, with white rubber liner
    33-430
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 239,00 €
    239,00 €
    1 * 72 ST
       239,00 €
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Black
    Screw caps, with white rubber liner
    38-430
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 335,00 €
    335,00 €
    1 * 72 ST
       335,00 €
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Black
    Screw caps, short skirted, with white rubber liner
    51-400
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 764,00 €
    764,00 €
    1 * 72 ST
       764,00 €