Mounting medium, Entellan® new
Supplier: Merck
Certificates
About this item
Entellan® new is a new mounting medium for cover slippers and is specially suited for commercial automated mounting instruments that operate with cover slippers. Its viscosity ranges between 250 and 600 mPas, 1,00869 has a small viscosity range. This narrow viscosity range improves and accelerates the application, as neither the dropping rate nor the volume need to be reset after the first run with a new bottle of Entellan® new.
- Easy application
- Small viscosity range, 500 to 600 mPas (1,00869)
- Glass bottle