Volumetric flasks, class A
Supplier: VITLAB
Jump to:
About this item
PMP, opaque, with NS stoppers, PP.
- With ring mark individually calibrated to ‘In’
- With printed lot number and batch certificate
- Thermal stress up to 121 °C (autoclaving) does not permanently exceed the tolerance limit
- To preserve markings, cleaning at no higher than 60 °C is recommended
UV-absorbing, highly transparent. For storage of light-sensitive substances.
Certifications: Class A tolerances according to DIN EN ISO 1042.
7 Options Available Below
Product Details & Documents
PMP, opaque, with NS stoppers, PP.
- With ring mark individually calibrated to ‘In’
- With printed lot number and batch certificate
- Thermal stress up to 121 °C (autoclaving) does not permanently exceed the tolerance limit
- To preserve markings, cleaning at no higher than 60 °C is recommended
UV-absorbing, highly transparent. For storage of light-sensitive substances.
Certifications: Class A tolerances according to DIN EN ISO 1042.
Product Family Options
Product Information
- CapacityNSHeightToleranceAvailabilityPrice