Orders LinkContactUsLinkComponent
Buckets, Round
Buckets, Round
Supplier:   VITLAB
Buckets, Round
Supplier:  VITLAB
Certificates
Buckets, Round
2 Options Available Below
Product Details & Documents
PP, transparent.
  • Graduated
  • Very good thermal resistance
  • Very good chemical resistance
Universal multipurpose bucket with spout, reinforced rim and handle.
Product Family Options
Product Information
  • Division
    Colour
    Capacity
    Ø
    Height
    Availability
    Price
  • 1000 ml
    Transparent
    12 L
    310 mm
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,80 €
    32,80 €
    1 * 1 ST
       32,80 €
  • 1000 ml
    Transparent
    15 L
    310 mm
    370 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,20 €
    36,20 €
    1 * 1 ST
       36,20 €