Order Entry
Austria
ContactUsLinkComponent
VWR® Q Path® Slide Organisers
VWR® Q Path® Slide Organisers
Catalog # 631-1049
Supplier:  VWR Collection
undefined
VWR® Q Path® Slide Organisers
Catalog # 631-1049
Supplier:  VWR Collection

Specifications

  • No. of slides:
    2700
  • Colour:
    Green
  • Description:
    Organiser for transparencies with 7 drawers
  • Material:
    Metal
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

This metallic cabinet is suitable for the storage of 50×50 mm or 80×57 mm microscopy slides.

Weight: 9 kg

Weight: 9 kg