ContactUsLinkComponent
WHEATON® OmniSpence® ELITE, WHEATON® UniSpense® PRO Peristaltic Pump
WHEATON® OmniSpence® ELITE, WHEATON® UniSpense® PRO Peristaltic Pump
Supplier:  DWK Life Sciences
Certificates
undefined
WHEATON® OmniSpence® ELITE, WHEATON® UniSpense® PRO Peristaltic Pump
Supplier:  DWK Life Sciences
Specifications
  • Catalog No:
  • 224-0104
  • 224-0103
  • Model:
  • UniSpense® PRO
  • OmniSpense® ELITE
  • Rotation speed:
  • 75 and 150 rpm
  • 0 - 400 rpm
  • Int. Ø tubing:
  • 2, 3, 6, 8 mm
  • 2, 3, 6, 8 mm
  • Dosing volume:
  • 0,01 - 999,99 ml
  • 0,01 - 999,99 ml