Order Entry
Austria
ContactUsLinkComponent
M13KO7 Helper Phage, Cytiva
M13KO7 Helper Phage, Cytiva
Catalog # 27-1524-01
Supplier:  Cytiva
CAS Number:  
undefined
M13KO7 Helper Phage, Cytiva
Catalog # 27-1524-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    M13KO7 helper phage

Specifications

About this item

M13 phage for the GST expression system.

  • For phage rescue
  • Colourless to light yellow liquid
  • Concentration >5×10¹º pfu/ml

The M13KO7 helper phage is an M13mp1 derivative containing a modified gene 11 (G changed to T at position 6,125, giving a met to ile change at codon 40 of the gene 11 protein), an insertion of the origin of replication of p15A and the kanamycin resistance gene from Tn903 at the Ava I site (position 5,825).