ContactUsLinkComponent
Polyribonucleotide poly(I)·poly(C)
Polyribonucleotide poly(I)·poly(C)
Catalog # 27-4732-01
Supplier:  Cytiva
CAS Number:  
undefined
Polyribonucleotide poly(I)·poly(C)
Catalog # 27-4732-01
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    265 mg
  • Description:
    Poly(I)·Poly(C)
Specifications
About this item
The double-stranded polyribonucleotide poly(I)·poly(C) induces interferon in rabbit cells, and there has been interest in its effectiveness for inducing human interferon in vitro.