ContactUsLinkComponent
Cell culture reagents, microcarriers, Cytopore 2
Cell culture reagents, microcarriers, Cytopore 2
Catalog # 17-1271-01
Supplier:  Cytiva
undefined
Cell culture reagents, microcarriers, Cytopore 2
Catalog # 17-1271-01
Supplier:  Cytiva
Specifications
  • Description:
    Cytopore 2, dry powder
  • Pk:
    20 g
Specifications
About this item
Cytopore 2 macroporous microcarriers are principally designed for use in suspension culture systems for growth of adherent diploid cells requiering a higher surface charge and the production of recombinant proteins.
  • Macroporous structure of Cytopore microcarriers encourages cell growth into the bead, while the micropores provide maximum nutrient availability
  • The characteristics enable growth of cells which require high recirculation rates and high nutrient-availability
  • Intended for culture of truly anchorage-dependent cells which require a very high charge-density for optimal growth
  • Transparent for easy microscopic examination of attached cells